LaMotte® Secchi Disk
Secchi Disk with Calibrated Line, 7.75"
470112-626
: 0171-CL
This 8", black and white disk is used to determine the turbidity of natural water.
- Black and White design
- Sounding lead and line included
- Function: to determine the turbidity of water
- Aligned with GLOBE protocols
The disk is raised or lowered several times to determine the average depth at which it disappears from sight.
: 470112-626 is complete set, containing a secchi disk and a calibrate line. 470004-364 is the standalone rope, and 470101-430 the standalone disk.
- Description:Secchi Disk with Calibrated Line
- O.D.:7.75"



