Edvotek® Variable Micropipettes
2 racks of 96 tips - Blue, 101 - 1000 µl
Catalog # 470105-436
Supplier: EDVOTEK, INC.
Supplier #: 637
Jump to:
Product Details & Documents
Our newly re-designed variable micropipettes are sturdily designed with volumes ranging from 0.1 to 5000 µl.
- New and improved
- High quality, same price
- More durable and ergonomic
- Lifetime warranty
- Light gray color
They are easy to use, highly accurate and use standard micropipette tips. The volume is easily selected by twisting the top. The lightweight design and tip ejector make operation fast and easy.
Ordering information: A tool and instructions are included for self-calibration.
Documents
Specifications
- Volume:101 - 1000 µl
- Description:2 racks of 96 tips - Blue