Order Entry
Mexico
Orders LinkContactUsLinkComponent
Rapitest Soil Test Kit
Test kit, soil pH, Rapitest, 10
  470104-846
Rapitest Soil Test Kit
  470104-846
 :  
Rapitest Soil Test Kit
Rapitest Soil Test Kit
Test kit, soil pH, Rapitest, 10
 :  
$15.46
Each (1UN) Case de 12 (1UN)
Perform qualitative soil tests without the use of chemical reagents and test tubes.
  • No hazardous compounds required for testing
  • Colorimetric determination of soil components
  • Instructions included
This kit lets students use a simple three-step process involving water, tablets and color comparators. To test soil, simply mix it with water and let it settle. Then pour the liquid off into the color comparator, add a tablet and compare the liquid with the color film chart for a test reading.
470104-846 includes 10 pH soil test kits and fully instructions.
470175-090 includes 10 tests each for pH, nitrogen, phosphorus and potash and full instructions.
  • Description:
    Test kit, soil pH, Rapitest
  • No. of Tests:
    10