Unmounted Mirrors
Convex mirror, 50 mm, 50 mm
Catalog # 470104-378
Supplier: UNITED SCIENTIFIC SUPPLIES
Supplier #: MCV502
Jump to:
Product Details & Documents
Converging and diverging mirrors for optical experiments.
- Optically true glass
- Scratch-resistant copper back silvering
An excellent selection of spherical, silvered glass mirrors for demonstrations and experiments in optics.
Specifications
- Description:Convex mirror
- Focal Length:50 mm
- Diameter:50 mm