Order Entry
Mexico
Orders LinkContactUsLinkComponent
Replacement Bulbs
Replacement Bulbs
Replacement Bulb, with Bayonet Base, 110 V, 15 W
Catalog # 470103-538
Supplier #:  1101002400482
Replacement Bulbs
Catalog # 470103-538
Replacement Bulbs
Replacement Bulbs
Replacement Bulb, with Bayonet Base, 110 V, 15 W
Price
$95.96
Log in for availability
Each (1UN)
Product Details & Documents
Replacement microscope light bulbs are available for a variety of microscopes. If unsure which bulb type is needed, please contact Ward's Science.
Specifications
  • Description:
    Replacement Bulb, with Bayonet Base
  • Electrical:
    110 V, 15 W