Order Entry
Mexico
Orders LinkContactUsLinkComponent
Urine Test Strips
Urine Test Strips
Urine Test Strips, 5 Parameters Pk/100
Catalog # 470101-700
Supplier:   LW SCIENTIFIC SE
Supplier #:  URS-05PR-7777
Urine Test Strips
Catalog # 470101-700
Supplier:  LW SCIENTIFIC SE
CAS Number:  
Urine Test Strips
Urine Test Strips
Urine Test Strips, 5 Parameters Pk/100
CAS Number:  
Price
$759.53
Log in for availability
Each (1UN)
Product Details & Documents
Easy-to-use test strips check for up to 10 parameters.
  • Easy to Use
5 Parameter Urine Reagent Strips: Glucose, Protein, pH, Blood, Ketone
10 Parameter Urine Reagent Strips: Glucose, Protein, pH, Blood, Ketone, bilirubin, nitrite, urobilinogen, specific gravity, and leukocytes.
Specifications
Specifications
  • Description:
    Urine Test Strips, 5 Parameters Pk/100
Specifications