Order Entry
Mexico
Orders LinkContactUsLinkComponent
Wire Mesh Screens

Wire Mesh Screens

Screen, Aluminum Mesh for Window 8×16", 10", 10"
Catalog # 470040-048
Supplier #:  .011WIRE
Wire Mesh Screens
Catalog # 470040-048
Wire Mesh Screens

Screen, Aluminum Mesh for Window 8×16", 10", 10"

Price
$69.30
Log in for availability
Each (1UN)
Product Details & Documents
Specifications
  • Description:
    Screen, Aluminum Mesh for Window 8×16"
  • Length:
    10"
  • Width:
    10"
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.