Order Entry
Mexico
Orders LinkContactUsLinkComponent
 

Cross Section Models

Cross Section Animal Cell, 2, 14 cm, 15 cm, 14 cm
  470039-530
 :  LER1900
Cross Section Models
  470039-530
Cross Section Models
Cross Section Models

Cross Section Animal Cell, 2, 14 cm, 15 cm, 14 cm

 
$699.30
   
Each (1UN)
 

Soft Models Are Great For Small Hands

  • Show Basic Structures
  • One Half Of Each Model Serves As The Key To The Other Half
  • Great For Toss Around Activities

These soft foam models split in half to reveal general structures labeled on one side and numbered on the other. These are great for students to quiz each other on basic structure, or to take home for self study. Models include an activity guide with basic facts about each structure.

 
  • Description:
    Cross Section Animal Cell
  • Height:
    14 cm
  • Length:
    15 cm
  • Width:
    14 cm
  • No. of Parts:
    2
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.