Foam Bones
Foot
Catalog # 470030-126
Supplier: APTIC SUPERBONES
Supplier #: 823802
Jump to:
Product Details & Documents
Economical And Anatomically Accurate
- Inexpensive Generalized Bone Forms
- Dissect, And Break Similar To Real Bone
Individual solid FoamBones are lightweight and durable. They are excellent for showing external surface details.
Specifications
- Description:Foot