Order Entry
Mexico
Orders LinkContactUsLinkComponent
Somso® Gorilla Skull

Somso® Gorilla Skull

Gorilla Skull, Male, 2, 18 cm, 34 cm, 16 cm
Catalog # 470029-318
Supplier:   MARCUS SOMMER
Supplier #:  ZOS 50
Somso® Gorilla Skull
Catalog # 470029-318
Supplier:  MARCUS SOMMER
Somso® Gorilla Skull
Somso® Gorilla Skull

Gorilla Skull, Male, 2, 18 cm, 34 cm, 16 cm

Price
$10,100.00
Log in for availability
Each (1UN)
Product Details & Documents

Compare the structural differences between male and female gorillas (Gorilla gorilla), noting the smaller overall size, and size of central crest and brow ridges, of the female, by using this unmounted plastic, adult female skull with the SOMSO male gorilla skull. Both feature a removable lower jaw.

  • Cast from real specimens
  • Male and female versions available for comparison
  • 5 year manufacturers warranty

Documents

Specifications
  • Description:
    Gorilla Skull, Male
  • Height:
    16 cm
  • Length:
    34 cm
  • Width:
    18 cm
  • No. of Parts:
    2
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.