Order Entry
Mexico
Orders LinkContactUsLinkComponent
Cells: Structure, Function and Processes Posters

Cells: Structure, Function and Processes Posters

Mitosis Poster, Laminated, 61 cm, 46 cm
Catalog # 470023-476
Supplier #:  03-PS05-6L
Cells: Structure, Function and Processes Posters
Catalog # 470023-476
Cells: Structure, Function and Processes Posters
Cells: Structure, Function and Processes Posters

Mitosis Poster, Laminated, 61 cm, 46 cm

Price
$585.77
Log in for availability
Each (1UN)
Product Details & Documents

Zoom in on the key topics in cell biology with these colorful and innovative posters.

  • Beautifully illustrated
  • Highly detailed

Labeled diagrams and descriptive text highlight important structures and processes. Guaranteed to draw attention, the ten laminated posters include: Animal Cell, Meiosis, Plant Cell, Bacteria Cell, Mitosis and Cytokinesis, Cellular Respiration, Active and Passive Transport, DNA and RNA, Photosynthesis and Cell Specialization. Set of 10.

Specifications
  • Width:
    46 cm
  • Description:
    Mitosis Poster
  • Length:
    61 cm
  • Finish:
    Laminated
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.