Order Entry
Mexico
Orders LinkContactUsLinkComponent
Dissection Placemats

Dissection Placemats

Crayfish Dissection Placemat, Polyvinyl, 40 cm, 24.7 cm
Catalog # 470023-354
Supplier:   RANACO CORP SE
Supplier #:  420.5020.1
Dissection Placemats
Catalog # 470023-354
Supplier:  RANACO CORP SE
Dissection Placemats
Dissection Placemats

Crayfish Dissection Placemat, Polyvinyl, 40 cm, 24.7 cm

Price
$291.03
Log in for availability
Each (1UN)
Product Details & Documents

Perfect Instructional Companion for Wet Lab Areas

  • Resistant to spills of any kind

Illustrations and descriptive text are printed right on the durable polyvinyl mat, so students can have specimen classification and information on the internal and external anatomy right on the lab bench with them while they are dissecting. Can be easily cleaned with a sponge or a paper towel, so students don’t have to worry about damaging a text book or other paper copies.

Specifications
  • Width:
    24.7 cm
  • Description:
    Crayfish Dissection Placemat
  • Length:
    40 cm
  • Finish:
    Polyvinyl