Order Entry
Mexico
Orders LinkContactUsLinkComponent
Rectangular Supports
Rectangular Supports
4×6", ⁵/₁₆", 18"
Catalog # 470019-526
Supplier #:  SSB4X6-CI
Rectangular Supports
Catalog # 470019-526
Rectangular Supports
Rectangular Supports
4×6", ⁵/₁₆", 18"
Price
$388.56
Log in for availability
Each (1UN)
Product Details & Documents
Tip-free stability for experiments.
  • Glassware can sit directly on base
  • Rods are zinc-plated steel
Heavy cast iron bases with black enamel finish. Includes a removable zinc-plated steel rod.
Specifications
  • Rod Length:
    18"
  • Base Dimensions:
    4×6"
  • Rod Diameter:
    ⁵/₁₆"