Cellophane
Cellophane Roll, Yellow, 20×100"
Catalog # 470019-316
Supplier: HYGLOSS PRODUCTS, INC.
Supplier #: 71008
Jump to:
Product Details & Documents
Use the various colors to demonstrate plants’ reaction to light waves.
- Made from recycled materials
Also useful for demonstrating the effects of mixing various colors of light.
Specifications
- Color:Yellow
- Description:Cellophane Roll
- Size:20×100"