Order Entry
Mexico
Orders LinkContactUsLinkComponent
Rectangular Glass Coverslips

Rectangular Glass Coverslips

Glass cover slips, rectangular, 40×24 mm, 0.15 mm, #1
Catalog # 470019-010
Supplier #:  94-2440
Rectangular Glass Coverslips
Catalog # 470019-010
Rectangular Glass Coverslips
Rectangular Glass Coverslips

Glass cover slips, rectangular, 40×24 mm, 0.15 mm, #1

Price
$87.42
Log in for availability
Paquete de 1
Product Details & Documents

Traditional rectangular coverslip style

  • Quality slide making supplies
  • Traditional coverslip type

Traditional style coverslips to use when making slides in the laboratory.

#1 Thickness: 0.15 mm thick. Approximately 75–100 coverslips per ounce.
#2 Thickness: 0.2 mm thick. Approximately 50-75 coverslips per ounce.

Specifications
  • Numbering:
    #1
  • Description:
    Glass cover slips, rectangular
  • L×W:
    40×24 mm
  • Thickness:
    0.15 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.