Order Entry
Mexico
Orders LinkContactUsLinkComponent
Rectangular Glass Coverslips
Glass cover slips, rectangular, 40×22 mm, 0.15 mm, #1
  470019-008
 :  94-2240
Rectangular Glass Coverslips
  470019-008
Rectangular Glass Coverslips
Rectangular Glass Coverslips
Glass cover slips, rectangular, 40×22 mm, 0.15 mm, #1
$87.42
Paquete de 1
Traditional rectangular coverslip style
  • Quality slide making supplies
  • Traditional coverslip type
Traditional style coverslips to use when making slides in the laboratory.
#1 Thickness: 0.15 mm thick. Approximately 75–100 coverslips per ounce.
#2 Thickness: 0.2 mm thick. Approximately 50-75 coverslips per ounce.
  • Numbering:
    #1
  • Description:
    Glass cover slips, rectangular
  • L×W:
    40×22 mm
  • Thickness:
    0.15 mm