Slab Easels
Slab Easels, Slab Easel
Catalog # 470018-778
Supplier: JULE-ART INC.
Supplier #: 36 A
Jump to:
Product Details & Documents
Safely prop up flat specimens such as agate or fossils in a thin matrix. The smaller easel supports slabs up to 1/4" thick, the larger easel easily accommodates thicker slabs and crystal plates.
Slab Easel Size: 3"L x 2 1/2"W x 2 1/4"H. Package of four.
Thick Slab Easel Size: 2 1/2"L x 2 1/4"W x 2 3/4"H. Package of four.
Specifications
- Description:Slab Easels, Slab Easel