Order Entry
Mexico
Orders LinkContactUsLinkComponent
Eisco® Premium Wimshurst Machine
Eisco® Premium Wimshurst Machine
Premium Wimshurst machine, 35×20×40 cm
Catalog # 470017-088
Supplier:   VWR International
Eisco® Premium Wimshurst Machine
Catalog # 470017-088
Supplier:  VWR International
Eisco® Premium Wimshurst Machine
Eisco® Premium Wimshurst Machine
Premium Wimshurst machine, 35×20×40 cm
Price
$3,944.25
Log in for availability
Each (1UN)
Product Details & Documents
The durable Wilmshurst Machine is made for years of use.
  • Ideal for Classroom Education
  • Plastic and Aluminum Sheet Plates Featured
  • Two Leyden Jars Act as Condensers
  • Sturdy Base Secures Machine
The Electrostatic Wimhurst Generator consists of two 9 3/4" dia. circular plastic plates, which are rotated in opposite directions by a drive belt mechanism. Includes two Leyden jar condensers made from Corning glass and aluminum foil. The unit is mounted on a sturdy ABC base. It stands 15" tall and generates up to 75,000 volts!
A belt drives two high-resistance plastic and aluminum sheet plates, which are supported in two rigid uprights, in the opposite direction.
Two Leyden jars, made from glass and aluminum foil, act as condensers. All items are anchored on a sturdy base.
Specifications
  • Description:
    Premium Wimshurst machine
  • Dimensions:
    35×20×40 cm