Box Magnifiers
Box Magnifier
470016-878
: 6455100
Contains and Magnifies Small Specimens
These clear plastic boxes feature a magnifying press-fit lid; simply place the specimen to be studied in the box, cover, and observe. They can also be used when permanently embedding small specimens in resin. Size: 4X magnifier 1 L×1 W×7/8H"; 3X magnifier 1 1/2 L×1 1/2 W×1 1/2 H".
- Magnification:3×
- Description:Box Magnifier