Box Magnifiers
Box Magnifier
Catalog # 470016-876
Supplier: SKYWILL FAR EAST INDUSTRIES
Supplier #: 6455000
Jump to:
Product Details & Documents
Contains and Magnifies Small Specimens
These clear plastic boxes feature a magnifying press-fit lid; simply place the specimen to be studied in the box, cover, and observe. They can also be used when permanently embedding small specimens in resin. Size: 4X magnifier 1 L×1 W×7/8H"; 3X magnifier 1 1/2 L×1 1/2 W×1 1/2 H".
Specifications
- Magnification:4×
- Description:Box Magnifier