Order Entry
Mexico
Orders LinkContactUsLinkComponent
Solid Balls for Physics Experiments
Solid Balls for Physics Experiments
Solid Nylon Ball, 19 mm, ¾"
Catalog # 470016-782
Supplier #:  92062
Solid Balls for Physics Experiments
Catalog # 470016-782
Solid Balls for Physics Experiments
Solid Balls for Physics Experiments
Solid Nylon Ball, 19 mm, ¾"
Clearance
$1.60
Sale
Valid until 31/12/2026
Log in for availability
Each (1UN)
Product Details & Documents
Drilled balls are used for pendulum bobs or impact experiments, while our other balls are suitable for collision apparatus and density/volume experiments.
Caution: WARNING: CHOKING HAZARD -Item contains a small ball. Not for children under 3 years.
Specifications
  • Description:
    Solid Nylon Ball, 19 mm
  • Diameter:
    ¾"