Order Entry
Mexico
Orders LinkContactUsLinkComponent
Friction Rods
Friction Rods
Solid Glass, Rounded
Catalog # 470016-220
Número de proveedor #:  FRSG12
Friction Rods
Catalog # 470016-220
Friction Rods
Friction Rods
Solid Glass, Rounded
Price
$121.54
Log in for availability
Each (1UN)
Product Details & Documents
For your Electrostatic Needs!
  • Excellent quality
Friction rods are used for producing static charges when rubbed against materials such as wool, flannel, silk, and fur.
Specifications
  • Description:
    Solid Glass, Rounded