Order Entry
Mexico
Orders LinkContactUsLinkComponent
 

Equal Volume Density Cube Sets

Density cube, set of 4
Catalog # 470014-710
 :  DCSET4
Equal Volume Density Cube Sets
Catalog # 470014-710
Equal Volume Density Cube Sets
Equal Volume Density Cube Sets

Density cube, set of 4

Price
$300.60
Log in for availability
Each (1UN)
Product Details & Documents

High quality density cubes for the classroom.

  • Ideal for density experiments and to explore the physical properties of different materials
  • Compare masses for equal volumes
  • Uniform, reliable dimensions
  • Activity manual/reference guide included

Compare the masses and densities of these uniform 1" cubes. Explore the principles of bouyancy, and test the physical properties of metals and non-metals. Each set includes a reference table showing density values of individual specimens.

 : Set of 4 includes acrylic, pine, nylon, and aluminum, packaged in a poly bag.

Set of 10 includes acrylic, oak, pine, rosewood, steel, aluminum, copper, brass, nylon, and PVC, packaged in a wooden storage box.

Set of 12 includes acrylic, oak, pine, poplar, steel, aluminum, copper, brass, nylon, PVC, lignum vitae, and polypropylene, packaged in a plastic box.

Specifications
  • Description:
    Density cube, set of 4