Order Entry
Mexico
Orders LinkContactUsLinkComponent
Spherical Mirrors

Spherical Mirrors

Convex mirror, –100 mm, 50 mm
Catalog # 470013-312
Supplier:   VWR International
Spherical Mirrors
Catalog # 470013-312
Supplier:  VWR International
Spherical Mirrors
Spherical Mirrors

Convex mirror, –100 mm, 50 mm

Price
$29.32
Log in for availability
Each (1UN)
Product Details & Documents

These wide range of mirrors are perfect for investigating real and virtual images.

  • Variety of Sizes and Focal Lengths
  • Circular
  • Polished Glass Construction
  • Durable
  • Back-Surface Silvered

The circular mirrors are available in diameters of 38 mm, 50 mm, and 75 mm.

They're back-surface silvered, made of polished glass, and painted for durability.

Specifications
  • Description:
    Convex mirror
  • Focal Length:
    –100 mm
  • Diameter:
    50 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.