Order Entry
Mexico
Orders LinkContactUsLinkComponent
Spherical Mirrors

Spherical Mirrors

Concave mirror, 200 mm, 50 mm
Catalog # 470013-308
Supplier:   VWR International
Spherical Mirrors
Catalog # 470013-308
Supplier:  VWR International
Spherical Mirrors
Spherical Mirrors

Concave mirror, 200 mm, 50 mm

Clearance
$0.90
Sale

Valid until 31/12/2026

Log in for availability
Each (1UN)
Product Details & Documents

These wide range of mirrors are perfect for investigating real and virtual images.

  • Variety of Sizes and Focal Lengths
  • Circular
  • Polished Glass Construction
  • Durable
  • Back-Surface Silvered

The circular mirrors are available in diameters of 38 mm, 50 mm, and 75 mm.

They're back-surface silvered, made of polished glass, and painted for durability.

Specifications
  • Description:
    Concave mirror
  • Focal Length:
    200 mm
  • Diameter:
    50 mm