Order Entry
Mexico
Orders LinkContactUsLinkComponent
Equilateral Acrylic Prisms
Equilateral Acrylic Prisms
Prism, Equilateral/Acrylic, 75 mm x 25 mm
Catalog # 470013-002
Supplier #:  PAE075
Equilateral Acrylic Prisms
Catalog # 470013-002
Equilateral Acrylic Prisms
Equilateral Acrylic Prisms
Prism, Equilateral/Acrylic, 75 mm x 25 mm
Price
$177.50
Log in for availability
Each (1UN)
Product Details & Documents
Demonstrate refraction and internal reflection.
  • Highly polished sides
  • High optical clarity
Selection of equilaterial and right angle acrylic prisms.
Specifications
  • Description:
    Prism, Equilateral/Acrylic, 75 mm x 25 mm