Assorted Wires
Insulated copper wire, 4 oz., 25 m, 22
Catalog # 470012-766
Supplier: ARCOR ELECTRONICS CO INC SE
Supplier #: 22C-0-SCIE
Jump to:
Product Details & Documents
Specifications
- Description:Insulated copper wire, 4 oz.
- Length:25 m
- Gauge:22
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



