Balls for Physics Experiments
Drilled Aluminum Ball, 25 mm, 1"
Catalog # 470007-146
Supplier: EISCO SCIENTIFIC LLC
Supplier #: ALBL25
Jump to:
Product Details & Documents
Ideal for Collision and Pendulum Demonstrations
Drilled balls for pendulum bobs or impact experiments.
Specifications
- Description:Drilled Aluminum Ball, 25 mm
- Diameter:1"
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



