Order Entry
Mexico
Orders LinkContactUsLinkComponent
Wooden Splints

Wooden Splints

Wooden Splints, 114 mm, 4 mm
Catalog # 470005-838
Supplier #:  WDS-1B-500
Wooden Splints
Catalog # 470005-838
Wooden Splints
Wooden Splints

Wooden Splints, 114 mm, 4 mm

Price
$174.84
Log in for availability
Paquete de 500
Product Details & Documents

Wooden splints are used for mixing and applying in a laboratory setting.

Specifications
  • Description:
    Wooden Splints
  • Height:
    114 mm
  • Width:
    4 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.