Student Dissecting Scissors
Dissecting Scissors, 115 mm (4.5"), Stainless steel, Sharp/Sharp
Catalog # 470005-458
Supplier: DUNRITE INSTRUMENTS SE
Supplier #: 140601
Jump to:
Product Details & Documents
Great for general purpose dissection.
Great For General Purpose Dissection. These stainless Dissecting scissors will give you many years of service. Available in regular or Fine point.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Specifications
- Description:Dissecting Scissors
- Length:115 mm (4.5")
- Material:Stainless steel
- Blade Type:Sharp/Sharp
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



