Plane Mirrors
Plane Mirrors, Size, mm: 100 x 100 x 2.5
Catalog # 470111-290
Supplier: ABRISA-USPG INDUST. GLASS SE
Supplier #: 1-980
Jump to:
Product Details & Documents
All Types and Sizes for All Optical Applications
- Front surface silvering eliminates double reflections
- All produce virtual images
These plane mirrors are rectangular. They are available in glass or shatterproof Plexiglas. Back surface silvered mirrors are less expensive, and ideal for general use. The silvered back surface is painted for durability. Front surface silvering eliminates double reflections at the unsilvered surface, giving a brighter, sharper image. However, these mirrors are more easily damaged by scratching.
Specifications
- Description:Plane Mirrors, Size, mm: 100 x 100 x 2.5