Order Entry
Mexico
Orders LinkContactUsLinkComponent
Replacement Bulbs

Replacement Bulbs

Replacement Bulb, with Bayonet Base, 110 V, 30 W
Catalog # 470021-696
Supplier:   WALTER PRODUCTS SE
Supplier #:  LBB6
Replacement Bulbs
Catalog # 470021-696
Supplier:  WALTER PRODUCTS SE
Replacement Bulbs
Replacement Bulbs

Replacement Bulb, with Bayonet Base, 110 V, 30 W

Price
$133.26
Log in for availability
Each (1UN)
Product Details & Documents

Replacement microscope light bulbs are available for a variety of microscopes. If unsure which bulb type is needed, please contact Ward's Science.

Specifications
  • Description:
    Replacement Bulb, with Bayonet Base
  • Electrical:
    110 V, 30 W
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.