Order Entry
Mexico
Orders LinkContactUsLinkComponent
Dissolved Oxygen
Dissolved Oxygen
Catalog # 470021-232
Dissolved Oxygen
Catalog # 470021-232
Supplier:  
Dissolved Oxygen
Dissolved Oxygen
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Enough materials for five groups working simultaneously.
  • Contains enough materials for a total of 25 tests
  • The kit will measure 0, 4, and 8 ppm
Provide your students with hands-on reinforcement of key concepts in water quality and water treatment with this test kit. Includes a teacher’s manual with lesson plans, background information, and reproducible student procedure and data sheets.