Order Entry
Mexico
Orders LinkContactUsLinkComponent
Nitrate-Nitrogen
Nitrate-Nitrogen
LaMotte® Water Test Kit, Nitrate-Nitrogen
Catalog # 470021-202
Supplier #:  3354-01
Nitrate-Nitrogen
Catalog # 470021-202
Nitrate-Nitrogen
Nitrate-Nitrogen
LaMotte® Water Test Kit, Nitrate-Nitrogen
Price
$2,552.04
Log in for availability
Kit de 1
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Pre-calibrated test tubes and labware give fast, accurate results in the classroom or in the lab.
  • Range: 0 to 15 ppm
  • Material for 50 tests
  • Compact, waterproof carrying case contains kit
  • Instructions included
  • Pre-calibrated test tubes
  • Aligned with GLOBE protocols
Specifications
  • Description:
    LaMotte® Water Test Kit, Nitrate-Nitrogen