Environmentally Friendly Thermometers
Environmentally Friendly Thermometers, 1 °C, Total Immersion, −20...+150 °C, 12"
Catalog # 470019-648
Supplier: THERMCO PRODUCTS INC SE
Supplier #: B150CYBLK
Jump to:
Product Details & Documents
Glass thermometers with partial or total immersion.
Specifications
- Description:Environmentally Friendly Thermometers, 1 °C, Total Immersion
- Length:12"
- Range:−20...+150 °C
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



