Order Entry
Mexico
Orders LinkContactUsLinkComponent
Specific Gravity Hydrometer
Specific Gravity Hydrometer
Specific Gravity Hydrometer, 1.00–1.20, 0.002 divisions
Catalog # 470019-566
Supplier:   G&W INSTRUMENTS
Supplier #:  A-1101
Specific Gravity Hydrometer
Catalog # 470019-566
Supplier:  G&W INSTRUMENTS
Specific Gravity Hydrometer
Specific Gravity Hydrometer
Specific Gravity Hydrometer, 1.00–1.20, 0.002 divisions
Price
$313.40
Log in for availability
Each (1UN)
Product Details & Documents
Use these hydrometers with all processes where specific gravity or density of liquids is an important control.
The design of this hydrometer ensures easy cleaning and a low likelihood of breakage. The clear markings allow accurate readings. Accuracy: ±0.001. Size: 12"L.
Specifications
  • Description:
    Specific Gravity Hydrometer, 1.00–1.20, 0.002 divisions