Order Entry
Mexico
Orders LinkContactUsLinkComponent
Right Angle Clamp Holders
Right Angle Clamp Holders
Right Angle Clamp Holder, for 3/4" dia. Rods
Catalog # 470019-548
Supplier:   HUMBOLDT MFG CO
Supplier #:  H-8320
Right Angle Clamp Holders
Catalog # 470019-548
Supplier:  HUMBOLDT MFG CO
Right Angle Clamp Holders
Right Angle Clamp Holders
Right Angle Clamp Holder, for 3/4" dia. Rods
Price
$327.81
Log in for availability
Each (1UN)
Product Details & Documents
The perpendicular V-shaped jaws of these clamp holders are desiged to support rods and clamp extensions at a 90° angle.
Holders feature thumbscrews at a 180° angle.
Specifications
  • Description:
    Right Angle Clamp Holder, for 3/4" dia. Rods