Order Entry
Mexico
Orders LinkContactUsLinkComponent
Rectangular Supports

Rectangular Supports

5×8", ⅜", 20"
Catalog # 470019-494
Supplier:   SHIVDIAL SUD & SONS
Supplier #:  CH12321/2
Rectangular Supports
Catalog # 470019-494
Rectangular Supports
Rectangular Supports

5×8", ⅜", 20"

Price
$287.65
Log in for availability
Each (1UN) Case de 6 (1UN)
Product Details & Documents

Tip-free stability for experiments.

  • Glassware can sit directly on base
  • Rods are zinc-plated steel

Heavy cast iron bases with black enamel finish. Includes a removable zinc-plated steel rod.

Specifications
  • Rod Length:
    20"
  • Base Dimensions:
    5×8"
  • Rod Diameter:
    ⅜"