Order Entry
Mexico
Orders LinkContactUsLinkComponent
Tissue Forceps With Teeth

Tissue Forceps With Teeth

Tissue Forceps, Stainless steel, 110 mm (4.5"), 2 x 3 Tooth
Catalog # 470018-964
Supplier #:  141015
Tissue Forceps With Teeth
Catalog # 470018-964
Tissue Forceps With Teeth
Tissue Forceps With Teeth

Tissue Forceps, Stainless steel, 110 mm (4.5"), 2 x 3 Tooth

Price
$45.82
Log in for availability
Each (1UN)
Product Details & Documents

A true no slip hold.

  • Interlocking teeth prevent slipping
  • Ridged handles for a confident hold
  • Stainless construction prevents chipping

For general dissection, these 1 × 2 teeth firmly grasp tissues, while the serrated handles provide nonslip control. Gillie's type with straight blunt tips.

Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.

Specifications
  • Description:
    Tissue Forceps
  • Length:
    110 mm (4.5")
  • Material:
    Stainless steel
  • Tip Style:
    2 x 3 Tooth
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.