Order Entry
Mexico
Orders LinkContactUsLinkComponent
Sharp-Point Surgical Scissors

Sharp-Point Surgical Scissors

Surgical Scissors, 165 mm (6.5"), Stainless steel, Sharp/Sharp
Catalog # 470018-942
Supplier #:  140982
Sharp-Point Surgical Scissors
Catalog # 470018-942
Sharp-Point Surgical Scissors
Sharp-Point Surgical Scissors

Surgical Scissors, 165 mm (6.5"), Stainless steel, Sharp/Sharp

Price
$69.30
Log in for availability
Each (1UN) Caja de 10 (1UN)
Product Details & Documents

Start your cuts easily.

  • Dissect without a scalpel
  • Handles rounded for ambidextrous use
  • Available in two sizes

Sharp-pointed, straight surgical scissors allow the user to insert blades into tissue to start the cut. Larger scissors provide greater leverage for tough cutting; shorter scissors for finer work. Stainless steel.

Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.

Specifications
  • Description:
    Surgical Scissors
  • Length:
    165 mm (6.5")
  • Material:
    Stainless steel
  • Blade Type:
    Sharp/Sharp
Accessories

Product Information

  • Catalog #
    Length
    Material
    Availability
    Price
  • Surgical Scissors
    Surgical Scissors
    470018-942
    165 mm (6.5")
    Stainless steel
    Log in for availability
    Each (1UN) - $69.30

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.