Order Entry
Mexico
Orders LinkContactUsLinkComponent
Easy Grip Fine-Point Forceps
Easy Grip Fine-Point Forceps
Easy Grip Fine-Point Forceps, Stainless steel, 101 mm (4"), Curved
Catalog # 470018-884
Supplier #:  140591
Easy Grip Fine-Point Forceps
Catalog # 470018-884
Easy Grip Fine-Point Forceps
Easy Grip Fine-Point Forceps
Easy Grip Fine-Point Forceps, Stainless steel, 101 mm (4"), Curved
Price
$97.01
Log in for availability
Each (1UN)
Product Details & Documents
Minimize hand fatigue.
  • Large gripping area
  • Fine and medium points available
  • Straight or curved tip selection
Comfortable to handle and incredibly easy to maneuver, these stainless steel forceps feature finger grips designed to prevent slipping and provide better control when working with specimens. Available in fine point, or medium point styles.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Specifications
  • Description:
    Easy Grip Fine-Point Forceps
  • Length:
    101 mm (4")
  • Material:
    Stainless steel
  • Tip Style:
    Curved