Science Take-Out® A Kidney Problem
A kidney problem unassembled 10 pack, 10
Catalog # 470017-794
Supplier: SCIENCE TAKE-OUT, LLC
Supplier #: 118U
Jump to:
Product Details & Documents
Does Your Patient Have Kidney Disease?
- Available As A Single Kit With Materials For 1 Student Or Group
- Unassembled 10 Pack For Additional Savings
- No Additional Equipment Needed
Analyze a simulated urine sample to determine if your patient's symptoms might be caused by kidney disease. Create a model to learn about normal kidney function. Propose an explanation for the patient's urine test results. This complete experiment setup contains all materials and does not require lab equipment.
Documents
Specifications
- Description:A kidney problem unassembled 10 pack
- No. of Lab Groups:10