Science Take-Out® Beebops Genetics And Evolution
Beebops genetics and evolution single kit, 1
Catalog # 470017-732
Supplier: SCIENCE TAKE-OUT, LLC
Supplier #: 105
Science Take-Out® Beebops Genetics And Evolution
Catalog # 470017-732
Supplier: SCIENCE TAKE-OUT, LLC
Jump to:
Product Details & Documents
Create A Baby Beebop!
- Available As A Single Kit With Materials For 1 Student Or Group
- Unassembled 10 Pack For Additional Savings
- No Additional Equipment Needed
Why are Beebop babies in the same family different? Model how sexual reproduction results in offspring with different characteristics. Which Beebop babies are most likely to survive and reproduce? Explore how natural selection affects the frequency of characteristics and genes in the Beebop population. This complete "dry lab" activity contains all required materials.
Documents
Manual Beebops.pdf
Manual
Specifications
- Description:Beebops genetics and evolution single kit
- No. of Lab Groups:1