Variable Speed Centrifuge
Centrifuge Adapters for 0.5-2.0mL Pediatric Tubes
Catalog # 470012-614
Supplier: UNITED PRODUCT & INSTRUMENT SE
Número de proveedor #: C800-04
Jump to:
Product Details & Documents
A Valuable Addition to Any Laboratory.
- 30 minute timer with bell tone signal
- Latching lid with Safety Switch
- Easily accessible stainless steel rotor
For a clean, complete separation of cellular components yielding a superior serum or plasma specimen, use this powerful variable speed centrifuge. Suction cup rubber feet for stability also reduce vibration. Centrifuge features all UL approved components.
Specifications
- Description:Centrifuge Adapters for 0.5-2.0mL Pediatric Tubes