Order Entry
Mexico
Orders LinkContactUsLinkComponent
Biocam Pathogen Charts

Biocam Pathogen Charts

Waterborne Pathogens Chart, Laminated, 61 cm, 45.7 cm
Catalog # 470006-420
Supplier #:  WC53
Biocam Pathogen Charts
Catalog # 470006-420
Biocam Pathogen Charts
Biocam Pathogen Charts

Waterborne Pathogens Chart, Laminated, 61 cm, 45.7 cm

Price
$802.17
Log in for availability
Each (1UN)
Product Details & Documents

These high-resolution microphotography charts will make an excellent addition to the walls of your classroom.

  • Provide superior definition of structures without the aid of a microscope
Specifications
  • Width:
    45.7 cm
  • Description:
    Waterborne Pathogens Chart
  • Length:
    61 cm
  • Finish:
    Laminated
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.