Foam Brushes
Foam Beaker Brush, Foam, Plastic
Catalog # 470004-702
Supplier: BRUSHTECH
Supplier #: B60C
Jump to:
Product Details & Documents
Foam brushes are designed for cleaning lab glass and plasticware.
- Brushes will not chip, scratch or mar glassware
- Handles are plastic, including grips
Brushes are made of a special foam that does not get wet, yet transports water and detergent to the glass surface. These brushes will easily out-live others. They should be a fixture in your prep room.
Specifications
- Bristle Material:Foam
- Handle Material:Plastic
- Description:Foam Beaker Brush