Order Entry
Mexico
Orders LinkContactUsLinkComponent
Foam Brushes
Foam Flask Brush, Foam, Plastic
  470004-700
 :   BRUSHTECH
 :  B55C
Foam Brushes
  470004-700
 :  BRUSHTECH
Foam Brushes
Foam Brushes
Foam Flask Brush, Foam, Plastic
$223.06
Each (1UN) Case de 48 (1UN)
Foam brushes are designed for cleaning lab glass and plasticware.
  • Brushes will not chip, scratch or mar glassware
  • Handles are plastic, including grips
Brushes are made of a special foam that does not get wet, yet transports water and detergent to the glass surface. These brushes will easily out-live others. They should be a fixture in your prep room.
  • Bristle Material:
    Foam
  • Handle Material:
    Plastic
  • Description:
    Foam Flask Brush