Student gratings
Student gratings, 600 lines per mm
Catalog # 470004-102
Supplier: UNITED SCIENTIFIC SUPPLIES
Supplier #: DFG600
Jump to:
Product Details & Documents
Quality gratings perfect for your Optics Needs
- Viewing area measures approx. 45x30mm
Quality transmission gratings protected by two glass plates, 60x45mm.
Specifications
- Description:Student gratings, 600 lines per mm