Order Entry
Mexico
Orders LinkContactUsLinkComponent
Hanging Cell Models

Hanging Cell Models

Plant Cell, 23, 1, 61 cm, 46 cm
Catalog # 470002-684
Supplier #:  B142
Hanging Cell Models
Catalog # 470002-684
Hanging Cell Models
Hanging Cell Models

Plant Cell, 23, 1, 61 cm, 46 cm

Price
$10,434.68
Log in for availability
Each (1UN)
Product Details & Documents

For Demonstration And Display

These high visibility large models are vacuum formed sheets of heavy plastic, molded to show in relief all the known organelles and inclusions of these two important cells. Twenty structures are included in the animal cell; twenty-three in the plant cell. All are identified by the accompanying key cards. Hand painted in brilliant color and are fitted with an easel back which permits the model to sit upright on a surface of hang on a wall with equal ease.

Specifications
  • Description:
    Plant Cell
  • Length:
    46 cm
  • No. of Keyed Items:
    23
  • Width:
    61 cm
  • No. of Parts:
    1