Surgical Scissors
Surgical Scissors, 110 mm (4.5"), Stainless steel, Sharp/Blunt
Catalog # 470001-842
Supplier: DUNRITE INSTRUMENTS SE
Supplier #: 140988
Jump to:
Product Details & Documents
Each feature a sharp point for penetration and a blunt point for situations where not desired.
Available in four sizes for most dissection needs.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Specifications
- Description:Surgical Scissors
- Length:110 mm (4.5")
- Material:Stainless steel
- Blade Type:Sharp/Blunt



